Learn More
Description
Specifications
Specifications
| Antigen | ZNF690 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | ZSCAN29/2610 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Accession No. | Q8IWY8 |
| Gene Alias | EC 2.1.1.43, EC 2.7.10.1, FLJ35867, MGC129894, MGC129895, Zfp690, zinc finger and SCAN domain containing 29, zinc finger and SCAN domain-containing protein 29, Zinc finger protein 690KOX31-like zinc finger protein, ZNF690 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
