Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF704 Antibody (PCRP-ZNF704-3C10), PerCP, Novus Biologicals™
SDP

Catalog No. NB187832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187832 Supplier Novus Biologicals Supplier No. NBP314168PCP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF704 Monoclonal antibody specifically detects ZNF704 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen ZNF704
Applications Flow Cytometry
Classification Monoclonal
Clone PCRP-ZNF704-3C10
Conjugate PerCP
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ16218, MGC10858, zinc finger protein 704
Host Species Mouse
Immunogen Recombinant full-length human ZNF704 protein (Uniprot: Q6ZNC4)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 619279
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.