Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF79 Antibody (OTI3D2), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274955V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274955V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274955V Supplier Novus Biologicals Supplier No. NBP274955V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF79 Monoclonal specifically detects ZNF79 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZNF79
Applications Western Blot
Classification Monoclonal
Clone OTI3D2
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias pT7, zinc finger protein 79, zinc finger protein 79 (pT7), ZNFpT7
Gene Symbols ZNF79
Host Species Mouse
Immunogen Full length recombinant protein of human ZNF79 (NP_009066) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7633.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.