Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF800 Antibody, Novus Biologicals™
SDP

Catalog No. NBP178199 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178199 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178199 Supplier Novus Biologicals Supplier No. NBP178199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF800 Polyclonal specifically detects ZNF800 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ZNF800
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_789784.2
Gene Alias zinc finger protein 800
Gene Symbols ZNF800
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 550 and 600 of human Zinc Finger Protein 800 using the numbering given in entry NP_789784.2 (GeneID 168850).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168850
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.