Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF804A Antibody, Novus Biologicals™
SDP

Catalog No. NBP243681 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243681 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243681 Supplier Novus Biologicals Supplier No. NBP243681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects ZNF804A in Human samples, and it is validated for Immunoprecipitation, Western Blotting

ZNF804A Polyclonal specifically detects ZNF804A in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ZNF804A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500
Formulation PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
Gene Alias C2orf10, ZNF804A zinc finger protein 804A
Gene Symbols ZNF804A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZNF804A. The exact sequence is proprietary.
Molecular Weight of Antigen 137 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91752
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.