Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF81 Antibody (PCRP-ZNF81-2C7), HRP, Novus Biologicals™
SDP

Catalog No. NB179260 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179260 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179260 Supplier Novus Biologicals Supplier No. NBP314081H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF81 Monoclonal antibody specifically detects ZNF81 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen ZNF81
Applications Western Blot, ELISA
Classification Monoclonal
Clone PCRP-ZNF81-2C7
Conjugate HRP
Formulation PBS
Gene Alias dJ54B20.6, FLJ44367, HFZ20X-linked 45, zinc finger protein 81, zinc finger protein 81 (HFZ20)
Host Species Mouse
Immunogen Recombinant full-length human ZNF81 protein (Uniprot: P51508)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347344
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.