Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF846 Antibody (ZNF846/2687), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB390113 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390113 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390113 Supplier Novus Biologicals Supplier No. NBP308846C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF846 Monoclonal antibody specifically detects ZNF846 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen ZNF846
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone ZNF846/2687
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias zinc finger protein 846
Host Species Mouse
Immunogen Recombinant full-length human ZNF846 protein (Uniprot: Q147U1)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162993
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.