Learn More
Description
Specifically detects IKZF4 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
Specifications
Specifications
| Antigen | IKZF4 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of human IKZF4. The exact sequence is proprietary. |
| Molecular Weight of Antigen | 64 kDa |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
