Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNHIT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179374
Description
ZNHIT1 Polyclonal specifically detects ZNHIT1 in Human samples. It is validated for Western Blot.Specifications
ZNHIT1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CG1I, CGBP1, Cyclin-G1-binding protein 1, H_DJ0747G18.14, p18 Hamlet, p18Hamlet, putative cyclin G1 interacting protein, zinc finger HIT domain-containing protein 1, Zinc finger protein subfamily 4A member 1, zinc finger protein, subfamily 4A (HIT domain containing), member 1, zinc finger, HIT domain containing 1, zinc finger, HIT type 1, zinc finger, HIT-type containing 1, ZNFN4A1 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 92%; Mouse: 85%; Zebrafish: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_006340 | |
ZNHIT1 | |
Synthetic peptide directed towards the N terminal of human ZNHIT1The immunogen for this antibody is ZNHIT1. Peptide sequence MVEKKTSVRSQDPGQRRVLDRAARQRRINRQLEALENDNFQDDPHAGLPQ. | |
100 μL | |
Apoptosis | |
10467 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction