Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZnT-8/SLC30A8 Antibody (815039), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB373889 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373889 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373889 Supplier Novus Biologicals Supplier No. FAB7936W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZnT-8/SLC30A8 Monoclonal antibody specifically detects ZnT-8/SLC30A8 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen ZnT-8/SLC30A8
Applications Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 815039
Conjugate DyLight 650
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias solute carrier family 30 (zinc transporter), member 8, Solute carrier family 30 member 8, zinc transporter 8, zinc transporter ZnT-8, ZNT8ZnT-8
Host Species Mouse
Immunogen E. coli-derived recombinant human ZnT-8, Lys268-Pro359, Accession # Q8IWU4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 169026
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.