Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZRANB1/Trabid Antibody, Novus Biologicals™
SDP

Catalog No. NB418586 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB418586 25 μL
NBP186918 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB418586 Supplier Novus Biologicals Supplier No. NBP18691825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZRANB1/Trabid Polyclonal specifically detects ZRANB1/Trabid in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZRANB1/Trabid
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias HRPE773, jacalin-like lectin domain containing 2, JCLN2, pancreatic adenocarcinoma upregulated factor, PAUF, PRO1567, zymogen granule protein 16 homolog B, zymogen granule protein 16 homolog B (rat)
Gene Symbols ZRANB1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:SSGNSQRRSPPATKRDSEVKMDFQRIELAGAVGSKEELEVDFKKLKQIKNRMKKTDWLFLNACVGVVEGDLAAIEAYKSSGGDIARQLT
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 54764
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.