Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZRANB1/Trabid Antibody, Novus Biologicals™
SDP

Catalog No. NBP101469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP101469 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP101469 Supplier Novus Biologicals Supplier No. NBP101469
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ZRANB1/Trabid Polyclonal specifically detects ZRANB1/Trabid in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen ZRANB1/Trabid
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2ug/ml, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q9UGI0
Gene Alias HRPE773, jacalin-like lectin domain containing 2, JCLN2, pancreatic adenocarcinoma upregulated factor, PAUF, PRO1567, zymogen granule protein 16 homolog B, zymogen granule protein 16 homolog B (rat)
Gene Symbols ZRANB1
Host Species Goat
Immunogen Peptide with sequence KWLDRYRQIRPCTS corresponding to C-Terminus according to NP_060050.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 54764
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.