Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZRSR2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP157317 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP157317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP157317 Supplier Novus Biologicals Supplier No. NBP157317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZRSR2 Polyclonal specifically detects ZRSR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZRSR2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q15696
Gene Alias CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2, Renal carcinoma antigen NY-REN-20, U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-relatedprotein 2, U2 small nuclear ribonucleoprotein auxiliary factor, small subunit 2, U2(RNU2) small nuclear RNA auxiliary factor 1-like 2MGC142014, U2AF1L2, U2AF1RS2MGC142040, U2AF1-RS2U2AF35-related protein, URPU2 small nuclear RNA auxiliary factor 1-like 2, zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2
Gene Symbols ZRSR2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ZRSR2 (zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2) The peptide sequence was selected from the C terminal of ZRSR2. Peptide sequence HHDDYYSRLRGRRNPSPDHSYKRNGESERKSSRHRGKKSHKRTSKSRERH The peptide sequence for this immunogen was taken from within the described region.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8233
Test Specificity This product is specific to Subunit or Isoform: -related protein 2.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.