Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSCAN2 Antibody (PCRP-ZSCAN2-1F8), Biotin, Novus Biologicals™
SDP

Catalog No. NB155734 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB155734 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB155734 Supplier Novus Biologicals Supplier No. NBP314151B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZSCAN2 Monoclonal antibody specifically detects ZSCAN2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen ZSCAN2
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-ZSCAN2-1F8
Conjugate Biotin
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias FLJ20595, zfp-29, ZFP29, zinc finger and SCAN domain containing 2, zinc finger and SCAN domain-containing protein 2, Zinc finger protein 29 homolog, Zinc finger protein 854, ZNF854zinc finger protein 29
Host Species Mouse
Immunogen Recombinant human fragment (aa 55-135) of ZSCAN 2 protein (Uniprot: Q7Z7L9)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54993
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.