Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSCAN21/ZFP38 Antibody (OTI2E5), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB274956A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274956A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274956A48 Supplier Novus Biologicals Supplier No. NBP274956AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZSCAN21/ZFP38 Monoclonal specifically detects ZSCAN21/ZFP38 in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ZSCAN21/ZFP38
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2E5
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp434L134, KOX25, NY-REN-21, Renal carcinoma antigen NY-REN-21, Zfp-38, ZFP38, zinc finger and SCAN domain containing 21, zinc finger and SCAN domain-containing protein 21, zinc finger protein 38, zinc finger protein 38 (KOX 25), Zinc finger protein 38 homolog, zinc finger protein NY-REN-21 antigen, Zipro1, ZNF38DKFZp686H10254
Gene Symbols ZSCAN21
Host Species Mouse
Immunogen Full length recombinant protein of human ZSCAN21/ZFP38 (NP_666019) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7589.0
Target Species Human, Mouse, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.