Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZSCAN30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552416
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA552416 100 μL
1 options

Catalog No. PIPA552416

Supplier: Invitrogen™ PA552416

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLPSQDRHFS LATFNRRIPT EHSVLESHES EGSFSMNSND ITQQSVDTRE KLYECFDCGK AFCQSSKLIR HQRIHTGERP YACKECGKAF SLSSDLVRHQ RIHSGE Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 64%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN30
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZSCAN30
Gene Accession No. Q86W11
Gene Alias zinc finger and SCAN domain containing 30; zinc finger and SCAN domain-containing protein 30; Zinc finger protein 397 opposite strand; zinc finger protein 397OS; ZNF397 opposite strand; ZNF397OS; ZNF917; ZNF-WYM; ZSCAN30
Gene Symbols ZSCAN30
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZSCAN30. Recombinant protein control fragment (Product #RP-89426).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100101467
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.