Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZUFSP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.00 - $487.50
Specifications
Antigen | ZUFSP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18039720
![]() |
Novus Biologicals
NBP18039720UL |
20 μL |
Each for $182.00
|
|
|||||
NBP180397
![]() |
Novus Biologicals
NBP180397 |
100 μL |
Each for $487.50
|
|
|||||
Description
ZUFSP Polyclonal specifically detects ZUFSP in Human samples. It is validated for Western Blot.Specifications
ZUFSP | |
Polyclonal | |
Rabbit | |
NP_659499 | |
221302 | |
Synthetic peptide directed towards the C terminal of human C6orf113. Peptide sequence LCLLILDPGCPSREMQKLLKQDIEASSLKQLRKSMGNLKHKQYQILAVEG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
C6orf113, chromosome 6 open reading frame 113, dJ412I7.3, zinc finger with UFM1-specific peptidase domain, zinc finger with UFM1-specific peptidase domain protein | |
ZUFSP | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title