Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZXDC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ZXDC |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17936820
![]() |
Novus Biologicals
NBP17936820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179368
![]() |
Novus Biologicals
NBP179368 |
100 μL |
Each for $487.50
|
|
|||||
Description
ZXDC Polyclonal specifically detects ZXDC in Human samples. It is validated for Western Blot.Specifications
ZXDC | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
dJ337O18.5, SWIM domain containing 1, zinc finger, SWIM-type containing 1 | |
ZXDC | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
NP_001035743 | |
79364 | |
Synthetic peptide directed towards the middle region of human ZXDCThe immunogen for this antibody is ZXDC. Peptide sequence NLKAHMKGHEQESLFKCEVCAERFPTHAKLSSHQRSHFEPERPYKCDFPG. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title