Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Zymosan, TLR2 Ligand
SDP

Catalog No. nbp226233 Shop All Novus Biologicals Products
Click to view available options
Quantity:
200 mg

Applications: Functional, In vitro assay, In vivo assay, Ligand Activation

Buffer: The Zymosan suspension solution is insoluble and remains opaque. Resuspend the solution by vortexing right before use. Dilution: Functional, In vitro assay, Ligand Activation Reconstitute with 50mg Zymosan with 1ml ethanol in a sterilized non-pyrogenic vial by vortexing to a final concentration of 50mg/ml. When diluting further, use sterilized endotoxin-free water or PBS

Specifications

Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Quantity 200 mg
For Use With (Application) This product is useful for activation of Toll-like Receptor 2 (TLR2). Stimulation of TLR2 has been reported with 1 to 10μL. Note: TLR2 activation at this range of concentrations of Zymosan has also been observed in our product validation tests. Use in In vivo assay and functional inhibition reported in scientific literature (PMID 25846753). Use in ligand activation reported in scientific literature (PMID 25957979). Use in in vitro reported in scientific literature (PMID 27351455).
Target Zymosan, TLR2 Ligand
Species Human
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.