Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ZZL 7

Click to view available options
Quantity:
10 mg
Description
ZZL-7 is a fast-acting antidepressant that enhances 5-HT (serotonin) signaling in forebrain circuits. ZZL-7 disrupts the interaction between the 5-HT transporter (SERT) and neuronal nitric oxide synthase (nNOS) in the dorsal raphe nucleus (DRN), which reduces extracellular 5-HT concentration and down-regulates 5-HT1A receptor auto-activity in the DRN. This reduced auto-inhibition enables increase in firing frequency of serotonergic neurons in the DRN and increases in 5-HT release in the medial hippocampus. ZZL-7 can readily cross the blood-brain barrier.
Specifications
Specifications
Chemical Name or Material | N-Acetyl-L-alanyl-L-valine methyl ester |
CAS | 99141-91-0 |
Quantity | 10 mg |
Target | SLC6A4/5-HTTLPR/Serotonin transporter |
Molecular Formula | C11H20N2O4 |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction