accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111762-30-2

SClMgBr

CAS RN 111762-30-2

IUPAC Name: 2-(bromomagnesio)-5-chlorothiophene
Synonyms: 2-(bromomagnesio)-5-chlorothiophene
Molecular Weight (g/mol): 221.78
Molecular Formula: C4H2BrClMgS
InChi Key: BTGXLAKETDWIAB-UHFFFAOYSA-M
SMILES: ClC1=CC=C([Mg]Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results