CAS RN 1243259-19-9
CAS RN 1243259-19-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ LM11A 31 dihydrochloride
Nonpeptide p75NTR ligand; neuroprotective