accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1354485-04-3

OOHOOOOHNH(S)

CAS RN 1354485-04-3

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[4-(4-hydroxyphenoxy)phenyl]propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[4-(4-hydroxyphenoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 495.53
Molecular Formula: C30H25NO6
InChi Key: LFDWCUHMDBIXPE-NDEPHWFRSA-N
SMILES: OC(=O)[C@H](CC1=CC=C(OC2=CC=C(O)C=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)