accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 136440-22-7

H3CNNOH3CCH3

CAS RN 136440-22-7

IUPAC Name: 2,2,4-trimethyl-2H-imidazol-1-ium-1-olate
Synonyms: 2,2,4-trimethylimidazol-1-ium-1-olate
Molecular Weight (g/mol): 126.16
Molecular Formula: C6H10N2O
InChi Key: CNIGTNPXWSLRPH-UHFFFAOYSA-N
SMILES: CC1=NC(C)(C)[N+]([O-])=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results