CAS RN 172889-26-8
CAS RN 172889-26-8
IUPAC Name:
1-tert-butyl-3-(4-methylphenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms:
1-tert-butyl-3-(4-methylphenyl)pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol):
281.36
Molecular Formula:
C16H19N5
InChi Key:
ZVPDNRVYHLRXLX-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C1=NN(C2=NC=NC(N)=C12)C(C)(C)C
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Src Inhibitor, PP1
The Src Inhibitor PP1, also referenced under CAS 172889-26-8, controls the biological activity of Src. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
2
Tocris Bioscience™ PP 1
CAS: 172889-26-8 Molecular Formula: C16H19N5