accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 181180-42-7

SOSOOFFF

CAS RN 181180-42-7

IUPAC Name: 3,6-dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
Synonyms: 3,6-dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 248.23
Molecular Formula: C6H7F3O3S2
InChi Key: ZMDCEXLCDOPKGH-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results