Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog No. AC439750010
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181180-42-7
Molecular Formula C6H7F3O3S2
Molecular Weight (g/mol) 248.24
InChI Key ZMDCEXLCDOPKGH-UHFFFAOYSA-N
Synonym 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-thiopyran, 2,3dihydro-4-trifluoromethylsulfonyloxy-6h-thiopyran, 3,6-dihydro-2h-thiopyran-4-yltrifluoromethanesulfonate, trifluoro-methanesulfonic acid 3,6-dihydro-2h-thiopyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-thiopyran-4-yl ester, methanesulfonic acid, trifluoro-, 3,6-dihydro-2h-thiopyran-4-yl ester
PubChem CID 22625751
IUPAC Name 3,6-dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
SMILES C1CSCC=C1OS(=O)(=O)C(F)(F)F

Specifications

Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 248.24
Percent Purity 97%
Chemical Name or Material 3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.