accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3OONNNHCHOOCH3

CAS RN 183319-69-9

IUPAC Name: N-(3-ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride
Synonyms: erlotinib hydrochloride
Molecular Weight (g/mol): 429.9
Molecular Formula: C22H24ClN3O4
InChi Key: GTTBEUCJPZQMDZ-UHFFFAOYSA-N
SMILES: Cl.COCCOC1=C(OCCOC)C=C2C(NC3=CC=CC(=C3)C#C)=NC=NC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)