CAS RN 276881-63-1
CAS RN 276881-63-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Chemicals Tyrphostin AG 537, Thermo Scientific™
CAS: 276881-63-1 Molecular Formula: C23H20N4O6 MDL Number: MFCD00236440 Synonym: Bis-Tyrphostin