CAS RN 285983-48-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ p38 MAP Kinase Inhibitor X, BIRB 796
The p38 MAP Kinase Inhibitor X, BIRB 796, also referenced under CAS 285983-48-4, controls the biological activity of p38 MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
2
Tocris Bioscience™ BIRB 796
High affinity and selective p38 kinase inhibitor