accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3513-03-9

CH3NOHNOOHONNOH2NNH2NHNH2HCl

CAS RN 3513-03-9

IUPAC Name: 6-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3-[3-amino-5-(N-methylcarbamimidamido)pentanamido]-3,6-dihydro-2H-pyran-2-carboxylic acid hydrochloride
Synonyms: blasticidin-S hydrochloride
Molecular Weight (g/mol): 458.9
Molecular Formula: C17H27ClN8O5
InChi Key: YQXYQOXRCNEATG-UHFFFAOYNA-N
SMILES: Cl.CN(CCC(N)CC(=O)NC1C=CC(OC1C(O)=O)N1C=CC(N)=NC1=O)C(N)=N
Physical Form 
  • (3)
  • (4)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Type 
  • (9)
Color 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (9)
  • (7)
  • (1)
  • (3)
  • (1)