accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3538-65-6

H3CONHNH2

CAS RN 3538-65-6

IUPAC Name: butanehydrazide
Synonyms: butryic acid hydrazide
Molecular Weight (g/mol): 102.14
Molecular Formula: C4H10N2O
InChi Key: FCCCRBDJBTVFSJ-UHFFFAOYSA-N
SMILES: CCCC(=O)NN
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results