accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4342-03-4

H3CNCH3HNNNNH2NO(E)

CAS RN 4342-03-4

IUPAC Name: (4E)-4-(3,3-dimethyltriazan-1-ylidene)-4H-imidazole-5-carboxamide
Synonyms: dacarbazine
Molecular Weight (g/mol): 182.19
Molecular Formula: C6H10N6O
InChi Key: OMJKFYKNWZZKTK-UXBLZVDNSA-N
SMILES: CN(C)N\N=C1\N=CN=C1C(N)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)