accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHNNN

CAS RN 446859-33-2

IUPAC Name: 2-[5-(6-methylpyridin-2-yl)-1H-pyrazol-4-yl]-1,5-naphthyridine
Synonyms: 2-[3-(6-methylpyridin-2-yl)-2H-pyrazol-4-yl]-1,5-naphthyridine
Molecular Weight (g/mol): 287.33
Molecular Formula: C17H13N5
InChi Key: LBPKYPYHDKKRFS-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=N1)C1=C(C=NN1)C1=CC=C2N=CC=CC2=N1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)