accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53487-52-8

CH3SOOOH3CCH

CAS RN 53487-52-8

IUPAC Name: but-3-yn-2-yl 4-methylbenzene-1-sulfonate
Synonyms: but-3-yn-2-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 224.27
Molecular Formula: C11H12O3S
InChi Key: XWRDBUBKBUYLQI-UHFFFAOYNA-N
SMILES: CC(OS(=O)(=O)C1=CC=C(C)C=C1)C#C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results