accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59709-57-8

HBrCH3HNOHOH

CAS RN 59709-57-8

IUPAC Name: hydrogen 1-methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol bromide
Synonyms: hydrogen (+)-salsolinol bromide
Molecular Weight (g/mol): 260.13
Molecular Formula: C10H14BrNO2
InChi Key: OGMGXKJQIOUTTB-UHFFFAOYNA-N
SMILES: [H+].[Br-].CC1NCCC2=CC(O)=C(O)C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results