Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Dihydroxy-1-methyl-1,2,3,4-tetrahydroisoquinoline hydrobromide, 98%, Thermo Scientific™

Catalog No. AC335800010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59709-57-8
Molecular Formula C10H14BrNO2
Molecular Weight (g/mol) 260.131
MDL Number MFCD00012741
InChI Key OGMGXKJQIOUTTB-UHFFFAOYSA-N
Synonym 1-methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol hydrobromide, +/--salsolinol, hydrobromide, 6,7-dihydroxy-1-methyl-1,2,3,4-tetrahydroisoquinoline hydrobromide, +-salsolinol hydrobromide, 1-methyl-6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline hydrobromide, rac salsolinol, hydrobromide, 1,2,3,4-tetrahydro-1-methyl-6,7-isoquinolinediolhydrobromide, salsolinol hydrobromide hplc, 1,2,3,4-tetrahydro-1-methyl-6,7-isoquinolinediol hydrobromide, 1-methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol, bromide
PubChem CID 2733730
IUPAC Name 1-methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrobromide
SMILES CC1C2=CC(=C(C=C2CCN1)O)O.Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 1 g
Beilstein 21, III, 2121
Formula Weight 260.13
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dihydroxy-1-methyl-1,2,3,4-tetrahydroisoquinoline hydrobromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.