accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NH3CNNH3CCH3

CAS RN 65-61-2

IUPAC Name: hydrogen N1,N1,N7,N7-tetramethylacridine-1,7-diamine chloride
Synonyms: acridine orange
Molecular Weight (g/mol): 301.82
Molecular Formula: C17H20ClN3
InChi Key: ASKSWIKIIFUEOI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)C1=CC=C2N=C3C=CC=C(N(C)C)C3=CC2=C1
Color 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)