CAS RN 83314-01-6
IUPAC Name:
(1S,5Z,7R,8E,11S,12S,13E,15S,21R)-25-(acetyloxy)-1,11,21-trihydroxy-17-[(1R)-1-hydroxyethyl]-5,13-bis(2-methoxy-2-oxoethylidene)-10,10,26,26-tetramethyl-19-oxo-18,27,28,29-tetraoxatetracyclo[21.3.1.1³,⁷.1¹¹,¹⁵]nonacos-8-en-12-yl (2E,4E)-octa-2,4-dienoate
Synonyms:
(1S,5Z,7R,8E,11S,12S,13E,15S,21R)-25-(acetyloxy)-1,11,21-trihydroxy-17-[(1R)-1-hydroxyethyl]-5,13-bis(2-methoxy-2-oxoethylidene)-10,10,26,26-tetramethyl-19-oxo-18,27,28,29-tetraoxatetracyclo[21.3.1.1³,⁷.1¹¹,¹⁵]nonacos-8-en-12-yl (2E,4E)-octa-2,4-dienoate
Molecular Weight (g/mol):
905.04
Molecular Formula:
C47H68O17
InChi Key:
MJQUEDHRCUIRLF-BXOMOELISA-N
SMILES:
CCC\C=C\C=C\C(=O)O[C@H]1\C(C[C@H]2CC(OC(=O)C[C@H](O)CC3CC(OC(C)=O)C(C)(C)[C@](O)(CC4C\C(C[C@@H](O4)\C=C\C(C)(C)[C@]1(O)O2)=C\C(=O)OC)O3)[C@@H](C)O)=C\C(=O)OC
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Bryostatin 1
A macrocyclic lactone with antitumor properties.
2
R&D Systems™ Bryostatin 1
CAS: 83314-01-6 Molecular Formula: C47H68O17