accessibility menu, dialog, popup

UserName

Viewing profile as user:
KSHSO

CAS RN 83373-60-8

IUPAC Name: tricyclo[5.2.1.0²,⁶]decan-8-yl sulfanylmethanethioate potassium
Synonyms: tricyclo[5.2.1.0²,⁶]decan-8-yl sulfanylmethanethioate potassium
Molecular Weight (g/mol): 267.47
Molecular Formula: C11H16KOS2
InChi Key: YFUFHKMDCZBHMY-UHFFFAOYNA-N
SMILES: [K].SC(=S)OC1CC2CC1C1CCCC21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results