Brushes
Tools consisting of bristles, hair, or wire set into a handle that are used to clean, scrub, or apply a liquid or powder to a surface; products are available in several sizes, and may be used for specific applications.
Some laboratory brushes are used for manual cleaning of laboratory glassware and plasticware. These typically have a handle with terminal, central or lateral bristles to effectively clean specific types of labware and vessels. Most laboratory brushes are autoclavable or washable for repeated use. They are sold individually or as sets with different dimensions to allow for effective cleaning of a wide range of vessel sizes. Brushes may be tapered or tipped with a cotton wad or fine bristle tuft for effective cleaning of tube bases.
Other brushes may be used to clean weighing pans of balances or for other specific tasks.
What are Laboratory Brushes Made of?
Brush handles can be made of wood, of durable plastics such as polypropylene, or of metals such as brass or stainless-steel wire. The bristles can be made from a range of materials depending on the brush type:
• Nylon bristles are tough, hard wearing, and resistant to aggressive chemicals
• Natural bristles (for example goat hair) are suitable for more delicate cleaning
• White silk bristles are suitable for general use
What Types of Laboratory Brushes are Available?
• General-purpose cleaning brushes
• Test tube brushes
• Beaker or bottle brushes
• Burette or pipette brushes
• Flask brushes, with a bent handle for easy access to awkward corners
• Measuring cylinder brushes
• Chromatography brushes for injector and detector ports
• Centrifuge brushes
• Sieve brushes
• Square tank brushes
- (1)
- (6)
- (2)
- (1)
- (1)
- (5)
- (38)
- (8)
- (2)
- (15)
- (1)
- (3)
- (7)
- (1)
- (3)
- (1)
- (82)
- (2)
- (1)
- (3)
- (1)
- (2)
- (1)
- (12)
- (1)
- (4)
- (2)
- (2)
- (4)
- (1)
- (2)
- (3)
- (1)
- (25)
- (2)
- (1)
- (1)
- (9)
- (1)
- (1)
- (3)
- (6)
- (2)
- (62)
- (1)
- (1)
- (5)
- (1)
- (4)
- (1)
- (79)
- (1)
- (3)
- (1)
- (22)
- (6)
- (2)
- (64)
- (17)
- (8)
- (6)
- (8)
- (5)
- (2)
- (21)
- (2)
- (1)
- (1)
- (4)
- (29)
- (293)
- (1)
- (77)
- (31)
- (3)
- (7)
- (1)
- (20)
- (2)
- (1)
- (5)
- (1)
- (1)
- (6)
- (6)
- (2)
- (10)
- (3)
- (11)
- (1)
- (1)
- (69)
- (1)
- (1)
- (8)
- (1)
- (1)
- (360)
- (1)
- (2)
- (2)
- (1)
- (5)
- (1)
- (1)
- (62)
- (377)
- (1)
- (2)
- (1)
- (28)
- (2)
- (1)
- (1)
- (5)
- (136)
- (12)
- (2)
- (4)
- (15)
- (70)
- (8)
- (15)
- (124)
- (9)
- (2)
Filtered Search Results
Fisherbrand™ Steel Wire Handled Brushes
Fisherbrand™ Steel Wire Handled Brushes feature stiff, white nylon bristles for cleaning test tubes.
| Tip Type | Radial Tufted End |
|---|---|
| Product Type | Test Tube Brush |
| Size | Small |
| Color | White |
| Bristle Material | Nylon |
| Bristle Type | Radial, Tufted End |
| For Use With (Application) | For cleaning test tubes |
| Handle Material | Galvanized steel wire |
Fisherbrand™ Budget-Priced Brushes
Fisherbrand™ Budget-Priced Brushes are an economical option for cleaning test tubes.
| For Use With (Equipment) | Test tubes |
|---|---|
| Tip Type | Radial Tufted End |
| Product Type | Test Tube Brush |
| Handle Material | Galvanized Wire |
Fisherbrand™ Anti-Static Static-Away™ Handle Brushes
Anti-Static Static-Away™ Handle Brushes designed with the softest hairs available to prevent scratching.
| Tip Type | Straight |
|---|---|
| Product Type | Anti-static Brush |
| Color-coded | No |
| Bristle Material | Synthetic Fibers |
| Bristle Type | Soft |
| Weight (English) | 0.5 oz. |
| Handle Type | Anti-static |
| Scent | Unscented |
| For Use With (Application) | Ideal for dusting balances, cuvets, microscope slides, optics and instrument/computer screens, designed with the softest hairs available to prevent scratching |
| Sterility | Non-sterile |
| Handle Material | Conductive Composite |
| Weight (Metric) | 14.17 g |
Fisherbrand™ Transfer Pipet Brush
Cone-shaped brush with stiff nylon bristles for cleaning transfer pipets.
Micronova™ M-Zone™ Bottle Brush
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Color-coded bottle brushes with rounded, stiff polyester bristles for versatile and aggressive cleaning of bottles and various equipment in production and in the lab.
| Disposable | Reusable |
|---|---|
| Content And Storage | Keep at ambient temperature |
| Size | 16 x 10 x 6 in. |
| Bristle Material | Polyester |
| Bristle Type | Soft |
| Quantity | 12/Cs. |
| Filtered | Non-filtered |
| Resistant To | Standard disinfectants and cleaning solutions |
| Material | Polypropylene, Polyester |
| Sterility | Non-sterile |
| Care Instructions | Keep at ambient temperature |
| Product Type | Bottle Brush |
| Certifications/Compliance | Certificate of Conformance; ISO 9001 Certified Facility; FDA Approved |
| Autoclavable | Not Autoclavable |
| Handle Material | Polypropylene |
Fisherbrand™ Heavy-Duty Brushes
Fisherbrand™ Heavy-Duty Brushes have spiral-wound bristles for cleaning beakers.
| Tip Type | Tuft End |
|---|---|
| Product Type | Heavy Duty Brush |
| Color | Black |
| For Use With (Application) | For cleaning beakers |
| Handle Material | Wood |
| No. of Bristle Rows | 1 |
|---|---|
| Includes | Hanging ring |
| Product Type | Flask Brush |
| Bristle Type | Soft |
| Handle Type | Flexible |
| Handle Material | Plastic |
Justman Brush Brush with Extension for Flask Curves
For cleaning bottom curves and inside shoulders of flasks and bottles