Clamps
Adjustable devices designed to securely hold two objects together or to hold an object in place; used with standard-taper and ball-and-socket joints, glass and tubing assemblies, and with ring stands. Available as plastic joint clips, stainless steel pinch clamps, and taper clips.
A laboratory clamp is an essential piece of equipment used to support and secure labware and other laboratory apparatus such as flasks, reaction vessels, tubing, thermometers, probes, columns, funnels, and overhead stirrers.
Some laboratory clamps consist of a shaft and boss head, secured with hand-tightened thumbscrews, knurled nuts, or wingnuts onto a supporting rod, stand or scaffold. They may also include two- or three-pronged, adjustable closing jaws designed for gripping objects safely and securely.
Laboratory clamps are constructed of metals such as stainless steel, nickel-plated zinc, plated brass or diecast aluminum alloy for enhanced chemical, heat, and corrosion resistance. However, some components such as the adjusting screws may be made of plastic, and the prongs may also be lined or covered with cork, plastic, glass-fiber, or rubber for enhanced grip with minimal damage or breakage.
Laboratory Clamp Styles and Designs
• Universal, general-purpose clamps that are fully adjustable for accommodating and securing a wide range of items of different sizes such as columns and thermometers
• Retort clamps for securing round-bottom flasks and similar glassware
• Fork clamps for securing and supporting cone-and-socket ground glassware joints
• Three-prong extension clamps, sometimes available with spring-loaded jaws, for holding irregularly shaped or outsized objects
• Specialized clamps for securing burettes and condensers or reaction vessels
• Chain clamps, which incorporate a circular, adjustable chain loop to hold cylinders and beakers
- (10)
- (3)
- (1)
- (7)
- (4)
- (1)
- (2)
- (28)
- (2)
- (1)
- (1)
- (5)
- (1)
- (8)
- (26)
- (2)
- (10)
- (1)
- (1)
- (8)
- (1)
- (30)
- (1)
- (1)
- (1)
- (1)
- (2)
- (1)
- (6)
- (1)
- (1)
- (4)
- (2)
- (2)
- (1)
- (1)
- (11)
- (1)
- (1)
- (1)
- (18)
- (1)
- (1)
- (1)
- (2)
- (1)
- (5)
- (1)
- (1)
- (1)
- (1)
- (1)
- (15)
- (1)
- (4)
- (11)
- (1)
- (1)
- (4)
- (2)
- (1)
- (1)
- (2)
- (2)
- (1)
- (1)
- (3)
- (4)
- (5)
- (2)
- (1)
- (2)
- (14)
- (2)
- (2)
- (3)
- (2)
- (1)
- (1)
- (1)
- (2)
- (2)
- (2)
- (2)
- (1)
- (2)
- (2)
- (3)
- (4)
- (4)
- (12)
- (1)
- (9)
- (1)
- (1)
- (7)
- (1)
- (1)
- (1)
- (3)
- (8)
- (3)
- (1)
- (1)
- (2)
- (10)
- (1)
- (5)
- (1)
- (1)
- (4)
- (10)
- (2)
- (3)
- (2)
- (12)
- (1)
- (1)
- (1)
- (6)
- (7)
- (1)
- (3)
- (14)
- (1)
- (2)
- (4)
- (12)
- (3)
- (2)
- (2)
- (2)
- (1)
- (1)
- (2)
- (1)
- (4)
- (1)
- (1)
- (1)
- (1)
- (19)
- (5)
- (9)
- (1)
- (1)
- (1)
- (9)
- (10)
- (1)
- (1)
- (20)
- (7)
- (17)
- (3)
- (1)
- (6)
- (2)
- (3)
- (1)
- (15)
- (1)
- (1)
- (9)
- (1)
- (1)
- (69)
- (1)
- (5)
- (2)
- (3)
- (14)
- (1)
- (1)
- (2)
- (1)
- (1)
- (7)
- (1)
- (16)
- (8)
- (1)
- (4)
- (2)
- (14)
- (37)
- (3)
- (2)
- (1)
- (6)
- (15)
- (7)
- (6)
- (5)
- (6)
- (6)
- (101)
- (2)
- (13)
- (6)
- (2)
- (21)
- (1)
- (11)
- (9)
- (30)
- (10)
- (1)
- (1)
- (9)
- (13)
- (3)
- (4)
- (2)
- (20)
- (1)
- (20)
- (1)
- (1)
- (1)
- (5)
- (1)
- (17)
- (19)
- (11)
- (1)
- (10)
Filtered Search Results
| Quantity | 1 Ea. |
|---|---|
| Material | Stainless Steel |
Troemner™ Talboys™ 3-Prong Dual Adjust Nickel-Plated Zinc Clamp
Designed to securely hold every type of laboratory glassware and apparatus. The Troemner™ 3-Prong Dual Adjust Nickel-Plated Zinc Clamp is constructed with round extension arms, which allow the clamps to be rotated 360°. Extension arms also allow placement of apparatus at various distances from lab-frames without compromising the integrity of your experiment.
Fisherbrand™ Fixed Position Medium Two-Prong Clamp
For holding apparatus near lab frame where no adjustment is required after set-up.
Fisherbrand™ Clamp, Tri-Grip
Clamp designed to securely hold every type of laboratory glassware and apparatus.
| Type | Mounting Clip, Reducer |
|---|---|
| Jaw Type | 3-Prong Dual Adjust |
| Color | Silver |
| Material | Zinc Alloy |
| Autoclavable | Autoclavable |
| Sterility | Non-sterile |
Fisherbrand™ Clamp, Tri-Grip, Micro
Tri-Grip Clamp allows irregular shapes to be held at any angle, surely and safely.
Troemner™ Talboys™ 3-Prong Dual Adjust Nickel-Plated Zinc Clamp
Designed to securely hold every type of laboratory glassware and apparatus. The Troemner™ 3-Prong Dual Adjust Nickel-Plated Zinc Clamp is constructed with round extension arms, which allow the clamps to be rotated 360°.
Fisherbrand™ Thermometer Swivel Clamp
For holding glass tubes and thermometers 114mm (4.49 in. ) from support rod.
Fisherbrand™ Polypropylene Clamp with Flow Control
Regulates flow through tubing from fully open to closed.
Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp.
Synthware™ Three Finger Adjustable Clamp, Small
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small, two-way adjustable, PVC coated prongs
Synthware™ Three Finger Adjustable Clamp, Large
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Large, two-way adjustable, PVC coated prongs.
| Type | Regular Clamp Holder |
|---|---|
| Size | Large |
| Size (English) Grip | 0.71 in. |
| Size (Metric) Grip | 1.8 cm |
| For Use With (Application) | Holding clamp to lab frames or ring stand |
Fisherbrand™ Clamp, Chain
Chain Clamp holds large round or irregular shaped objects firmly, yet gently to lab-frames and rods