Analyzers and Readers
Analyzers and Readers
Find the high-performance equipment you need to detect a variety of infections rapidly and accurately. A range of molecular analyzers and platforms can help you diagnose influenza A and B, RSV, strep A, and COVID-19. Our selection of clinical analyzers includes camera-based benchtop instruments, rapid chromatography immunoassay analyzers, and instruments offering microfluidic technology that deliver results in minutes.
Information About Molecular Analyzers
Molecular analyzers, also known as molecular diagnostic devices or molecular testing platforms, are advanced tools used to detect and identify specific genetic materials (such as DNA or RNA) of pathogens like viruses or bacteria. They play a crucial role in diagnosing respiratory illnesses, especially during outbreaks or pandemics, where rapid and accurate identification of pathogens is essential for timely patient management and public health interventions. Following is more information about molecular analyzers offered by the Fisher Healthcare brand.
Abbott ID NOW System (formerly Alere i)
The Abbottâ„¢ ID NOWâ„¢ System uses isothermal nucleic acid amplification technology to detect influenza A and B viral nucleic acids. The CLIA-waived methods include all the necessary testing components, and the analyzer offers barcode technology and LIS connectivity. Assays available for this platform include influenza A and B, RSV, strep A, and COVID-19.
Meridian Bioscience Revogene Instrument
Capable of single analyte or multiplex testing, the Meridian Bioscienceâ„¢ Revogeneâ„¢ molecular platform can help you standardize testing. Its fully automated molecular platform offers minimal hands-on time in a flexible, scalable platform that can be expanded to meet your changing needs. Performs homogenization, microorganism lysis, dilution, amplification, and detection of target nucleic acid sequences from multiple specimens using fluorescence-based real-time PCR. Assays are available for SARS-CoV-2, strep A, and several other non-respiratory infections.
QuidelOrtho Solana Molecular Platform
The QuidelOrthoâ„¢ Solanaâ„¢ Molecular Platform provides influenza A and B results in 45 minutes and performs up to 12 tests per run. The analyzer includes access to the QuidelOrtho proprietary wireless data management and surveillance system. Other CLIA-moderate assays for bacterial infections and infectious diseases are available for the Solana platform.
Information About Clinical Analyzers and Readers
Clinical analyzers help detect and measure various parameters, biomarkers, and analytes in patient samples. They are essential in diagnosing respiratory illnesses because they provide valuable information about the patient's health status and aid healthcare professionals in making accurate and timely treatment decisions. Following is more information about clinical analyzers and readers offered by the Fisher Healthcare brand.
Abbott DIGIVAL Reader
The Abbottâ„¢ DIGIVALâ„¢ Reader is a camera-based benchtop instrument that can objectively interpret results by using a camera to detect the presence of influenza types A and B and identify a completed lateral flow assay, analyze the intensity of the test and control line, and display the results (positive, negative, or invalid) on the color touchscreen.
BD Veritor Plus System
The BD Veritorâ„¢ Plus System is a rapid chromatography immunoassay analyzer for the qualitative detection of influenza A and B viral nucleoprotein antigens. The system is CLIA-waived for nasal and nasopharyngeal swabs and offers two modes: Analyze Now and Walk Away. This multi-assay platform includes tests for influenza A and B, RSV, and strep A.
LumiraDx Instrument
Innovative microfluidic technology delivers results in minutes when used with LumiraDx SARS-CoV-2 Ag Test strips. Authorized by FDA under EUA for use by laboratories certified under the CLIA, 42 U.S.C. §263a.
QuidelOrtho Sofia 2 Analyzer
The QuidelOrthoâ„¢ Sofiaâ„¢ 2 Analyzer accurately detects influenza A and B from nasal swabs, nasopharyngeal swabs, and nasal aspirate/wash samples. Its CLIA-waived procedure can produce dual test results from a single sample. The instrument features an easy-to-use interface with customizable settings.
Sekisui Diagnostics Acucy Reader
The Sekisui Diagnostics Acucyâ„¢ Reader System provides standardized test results in 15 minutes for CLIA-waived procedures. Use with the Sekisui Influenza A&B Test Kit.
  More
Featured Products
Molecular Analyzers
    Catalog Number Description Price Quantity & Availability  
Success
23-029-057 Details
Meridian Bioscience Revogeneâ„¢ SARS-CoV-2 Detectable Analytes: SARS-CoV-2
EDGE
Add to Cart
Product Specifications
For Use With (Equipment) Revogene Instrument (23-029-051)
Analysis Time 47 min. early call feature/85 min. result
Clia Complexity Moderate Complexity
DoA Calibrators No
Quantity 24 Tests/kit
Detection Method RT-PCR
Sample Type Nasopharyngeal, Oropharyngeal. Anterior Nasal, Mid-Turbinate Nasal Swab Sample
For Use With (Application) Nasopharyngeal, Oropharyngeal, Anterior Nasal, and Mid-Turbinate Nasal Swab Specimens
Disposable Single-use
Type COVID Test Kit
High-throughput Compatibility 8 Samples/Run
Sterility Sterile
Detectable Analytes SARS-CoV-2
CE Marker No
Includes 24 Disposable Transfer Tools, 24 Individual Pouches, Each Containing Revogene SARS CoV-2 Microfluidic Cartridge (PIE)
Form Ready-to-Use (RTU)
23-029-055 Details
Meridian Bioscienceâ„¢ Revogeneâ„¢ Strep A
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Includes One Sample Buffer Tube (SBT); One Disposable Transfer Tool (DTT); One GBS LB PIE
Quantity 24
Type Group A
Content And Storage 2°C to 25°C
23-029-056 Details
GSA_VA
Meridian Bioscienceâ„¢ Revogeneâ„¢ Carba C Quantity: 24
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Includes One Sample Buffer Tube (SBT); One Disposable Transfer Tool (DTT); One GBS LB PIE
Quantity 24
Type Colony
Content And Storage 2°C to 25°C
23-029-053 Details
GSA_VA
Meridian Bioscienceâ„¢ Revogeneâ„¢ GBS LB
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Includes One Sample Buffer Tube (SBT); One Disposable Transfer Tool (DTT); One GBS LB PIE
Quantity 24
Type Group B (Streptococcus)
Content And Storage 2°C to 25°C
23-029-054 Details
Meridian Bioscienceâ„¢ Revogeneâ„¢ C. difficile
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Includes One Sample Buffer Tube (SBT); One Disposable Transfer Tool (DTT); One GBS LB PIE
23-029-051 Details
GSA_VA
Meridian Bioscienceâ„¢ Revogeneâ„¢ Instrument
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Weight 10 kg
Voltage 110 to 240 V
Depth (Metric) 24 cm
Wattage 650 W
Width (Metric) 40 cm
23-044-180 Details
SDP
Quidel Solana SARS-CoV-2 LYO Assay Includes: Starter Kit- 48 Tests, 50 UTM Swab/ Analyzer
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Includes Starter Kit- 48 Tests, 50 UTM Swab/ Analyzer
Clia Complexity Moderate
Format Tube
For Use With (Application) RT-PCR, Rapid PCR
Disposable Single-use
Sterility Non-sterile
Detectable Analytes SARS-CoV-2
CE Marker CE-IVD
23-043-632 Details
Abbottâ„¢ ID NOW RSV Test Kit Quantity: 24/Pk.
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Clia Complexity Waived
Sensitivity Direct NPS: 98.6%, NPS eluted in VTM: 98.6%
Quantity 24/Pk.
Sample Type Nasopharyngeal Swab, Nasopharyngeal swab eluted in Viral Transport Media (VTM)
For Use With (Application) For diagnosis of Respiratory Syncytial Virus (RSV)
Specificity Direct NPS: 98%/NPS eluted in VTM: 97.8%
Type Test Kit
Test Time 13 min. or less
Content And Storage Room Temperature
Material Cardboard
Certifications/Compliance CLIA waived
Detectable Analytes Respiratory Syncytial Virus (RSV)
CE Marker Yes
Packaging Type Cardboard Box
State at Room Temperature Stable: Box 1 - powder Box 2 - Liquid
CPT Code 87634-QW
23-043-150 Details
GSA_VASDP
Quidel Molecular Strep A+G Control Set
Add to Cart
Product Specifications
Includes Control Kit (2 vials)
For Use With (Application)  Solanaâ„¢ GAS (25μL), Solana Step Complete (25μL) and Lyra Direct Strep (25μL).
Content And Storage 2°C to 8°C
23-043-123 Details
SDP
Quidel Solana RSV+hMPV Assay Kit Quantity: 48/Pk.
Due to product restrictions, please to purchase or view availability for this product.
EDGE
Product Specifications
Includes Process Buffer 48 tubes/kit,Reaction Tubes 48 tubes/kit
Quantity 48/Pk.
Sample Type Nasal swabs and nasopharyngeal swabs in transport media (BD/Copan VTM, Remel M4, Remel M4RT, Remel M
Sensitivity Drift BD/Copan VTM, Remel M4, Remel M4RT, Remel M5, Remel M6, or Copen eSwab
Content And Storage 2°C to 8°C
23-044-179 Details
SDP
Quidel Solana SARS-CoV-2 LYO Assay Includes: 48 Tests, 50 UTM/NS swab
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Includes 48 Tests, 50 UTM/NS swab
Clia Complexity Moderate
Format Tube
For Use With (Application) RT-PCR, Rapid PCR
Disposable Single-use
Sterility Non-sterile
Detectable Analytes SARS-CoV-2
CE Marker CE-IVD
23-043-107 Details
GSA_VASDP
Quidel Solanaâ„¢ Flu Assay Startup Kit Type: Influenza A and B Assay Kit
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Clia Complexity Moderate
Sensitivity 98.6.%/100.0% (Culture), 97.2%/100% (PCR)
For Use With (Application) For diagnosis of Influenza A&B
Quantity 48/Pk.
Specificity 95.1.% / 99.3% (culture)  96.8%/98.9% (PCR)
Type Influenza A and B Assay Kit
Test Time ≈45 min.
Content And Storage 2°C to 8°C
Control Sets M106
Detectable Analytes Influenza A and B
CE Marker Yes
Includes Process Buffer, Reaction Tubes
CPT Code 87502
Clinical Analyzers and Readers
    Catalog Number Description Price Quantity & Availability  
Success
23-043-083 Details
SDP
SOFIA STREP A WVD 25TST/PK RX
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Includes Cassettes, reagent tubes, reagent solution (contains Sodium Nitrite and Acetic Acid), reagent dropper tips, sterile rayon throat swabs, positive control swab and negative control swab
Quantity 25 Test Kit
Sample Type Throat Swab
Type Streptococcus A FIA (CLIA-Waived)
Test Time 5 min.
Content And Storage Room Temperature
Detectable Analytes Streptococcal Antigen (Group A)
23-043-151 Details
SDP
SARSANTGN FIA NAS SW 25T/PK RX
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
For Use With (Equipment) For use with Quidel Sofia 2 Analyzer, Cat. No. 20299.
Includes Cassettes, Reagent tubes, Reagent solution, Swabs, Pipettes (sufficient for 25 tests); Control swabs 1-pos 1-neg
Clia Complexity Waived
Description Test Kit
Quantity 25 Tests/Pk. or 300 Tests/Cs.
Sample Type Direct or VTM: Nasal swab, nasopharyngeal swab
Test Time 15 min.
Control Sets Positive and negative, included in kit
23-043-172 Details
SDP
Quidel Sofiaâ„¢ Strep A FIA CLIA-Waived Quantity: 25 Test Kit
This item has been discontinued by the supplier. Please to view product availability in your area.
Product Specifications
Includes Contents: 3 kits with 25 tests per kit; Includes: leased analyzer
For Use With (Application) Influenza testing
Type Sofia Flu Starter Kit
21-564-100 Details
GSA_VASDP
Quidel Sofiaâ„¢ 2 SARS Antigen Fluorescent Immunoassay (FIA) Sample Type: Direct or VTM: Nasal swab, nasopharyngeal swab
Add to Cart
Product Specifications
Includes Cassettes, reagent tubes, reagent solution (contains Sodium Nitrite and Acetic Acid), reagent dropper tips, sterile rayon throat swabs, positive control swab and negative control swab
Quantity 25 Tests
Sample Type Throat Swab
Type Streptococcus A FIA
Test Time 5 min.
Content And Storage Room Temperature
Detectable Analytes Streptococcal Antigen (Group A)
B256045 Details
Quidel Sofia Flu Starter Kit Includes: Contents: 3 kits with 25 tests per kit; Includes: leased analyzer
Due to product restrictions, please to purchase or view availability for this product.
EDGE
Product Specifications
Includes Lateral flow test strips, pipettes, Flu A+/B- control swab, Flu B+/A- control swab, RV reagent C in tubes
Clia Complexity Waived
Description Flu A + B Test Kit (For use in point of care settings)
Quantity 30 Assays
For Use With (Application) Detection of Flu A + B
Sample Type Nasal swabs and nasopharyngeal swabs
Type Flu A + B Test Kit
Test Time 10 min.
Detectable Analytes Influenza A and B
Packaging Type 1 Kit
23-044-175 Details
SDP
Quidel Sofiaâ„¢ Strep A FIA Quantity: 25 Tests
Add to Cart
B256049 Details
VER CTL SWB /-STRP A10PRPK
Add to Cart
Product Specifications
Quantity 10 Pairs
Type Control Swab
Sample Size Group A Streptococcus
23-044-176 Details
SDP
BD Veritorâ„¢ System Clinical Flu A+B Kit Quantity: 30 Assays
Add to Cart
23-046-682 Details
Quidel Corporation SOFIA2 CAMPY FIA 25T/PK
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Clia Complexity Waived
Description BinaxNOWâ„¢ Influenza A and B Card 2
Format Card
Quantity 22
Sample Type Nasal and NP
For Use With (Application) Influenza testing
Type Lateral Flow Assay
Test Time 15 min.
Content And Storage Room Temperature
Detectable Analytes Influenza
CE Marker No
Packaging Type Box
Formulation LF test card
Includes Controls, collection swabs
Shop by Brand