UserName
Viewing profile as user:
Epredia Digital Pathology Solutions
Epredia banner
Get Faster, More Accurate Results with Greater Ease
Experience outstanding speed, flexibility, and image quality for most sample types, empowering you to automate digital workflows from slide digitization through sign out.
The Epredia™ E1000 Dx Digital Pathology Solution is a fully integrated system designed to help laboratories achieve higher efficiency and image quality for their digital pathology workflows. With advanced capabilities spanning from digitization to case sign-out, the E1000 Dx automates and streamlines your processes for improved outcomes. Features include:
  • Automated QC rescan helps deliver the needed images to the pathologist
  • Dual slide processing to reduce downtime between scans and accelerate operations
  • AI-driven sample detection for optimized file size and higher throughput
  • Safety container to help ensure a single problematic slide does not stall the entire workflow
  • Large-capacity scanner holds up to 1,200 standard-width slides at once
  • Compatibility with Sakura™ and Leica coverslipper baskets provides flexibility for lab integration
Epredia E1000 Dx Digital Pathology Solution
Upgrade your whole-slide scanning capabilities with advanced efficiency and precision.
Epredia E1000 Dx Digital Pathology Solution
Prepare your laboratory for the future with 3DHISTECH™ and Epredia™ digital pathology solutions, a comprehensive range of digital slide scanners, software, and tissue microarrayers.
Pannoramic 1000
Free up your laboratory’s resources with the Pannoramic 1000 high-throughput whole-slide scanning solution.
Pannoramic 1000
Pannoramic 480
A whole-slide digital scanner designed for labs that require reliable and automated high-throughput scanning while maintaining image quality and flexibility.
Pannoramic 480
Pannoramic 250
An all-in-one solution for routine digital pathology research and storage. Enjoy increased speed with the ability to process 60 slides per hour.
Pannoramic 250
Pannoramic SCAN II
Save time in your routine pathology workflows with both brightfield and fluorescent scanning capabilities in the same machine.
Pannoramic SCAN II
Pannoramic MIDI II
A versatile, cost-effective digital pathology scanner with a feature set for low-volume scanning, making it a good fit for smaller labs.
Pannoramic MIDI II
Pannoramic DESK II
An entry-level scanner that features double-width scanning capability and works well for teleconsultation and remote section scanning.
Pannoramic DESK II
TMA Grand Master Automated Tissue Microarrayer
A fast, high-capacity tissue microarrayer that allows the precise selection and extraction of specific regions of interest on the donor blocks and their insertion into recipient blocks.
TMA Grand Master Automated Tissue Microarrayer
Reach Out to a Representative
Fill out the form and a Fisher Healthcare representative will contact you to answer your questions about Epredia Digital Pathology Solutions.
Required Field*
Step One: Account Information
Step Two: Company Information
State
Alaska
Alabama
Arkansas
Arizona
California
Colorado
Connecticut
D.C.
Florida
Georgia
Guam
Hawaii
Iowa
Idaho
Illinois
Indiana
Kansas
Kentucky
Louisiana
Massachusetts
Maryland
Maine
Michigan
Minnesota
Missouri
Mississippi
Montana
North Carolina
North Dakota
Nebraska
New Hampshire
New Jersey
New Mexico
Nevada
New York
Ohio
Oklahoma
Oregon
Pennsylvania
Puerto Rico
Palau
Rhode Island
South Carolina
South Dakota
Tennessee
Texas
Utah
Virginia
Virgin Islands
Vermont
Washington
Wisconsin
West Virginia
Wyoming
Military Americas
Military Europe/ME/Canada
Military Pacific
Country
United States
Canada
Step Three: Additional Information
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.