UserName
Viewing profile as user:
Rapid Critical Care Solutions
Rapid Critical Care Solutions
In the ED, you must identify life-threatening conditions and initiate appropriate treatment protocols for your patients. That’s why it’s so important to control costs and maximize workflow solutions for your department. To support those goals, the Fisher Healthcare channel has partnered with suppliers to offer a suite of high-quality and cost-effective solutions to improve clinical, operational, and financial outcomes for your facility.
Resources
Filename
fisher-healthcare-rapid-critical-care-solutions-brochure.pdf
Size
4 MB
Format
application/pdf
Siemens Healthineers epoc Blood Analysis System
Device for Testing Blood Gas, Electrolytes, and Metabolites 
Reduce the cost of testing and increase efficiency with the Siemens Healthineers epoc™ Blood Analysis System. This handheld device uses a single arterial, venous, or capillary whole blood sample to produce 13 critical blood gas, electrolyte, or metabolite test results in less than one minute. The barcoded test cards can be stored at room temperature and are automatically recognized and calibrated before the blood sample is introduced. Results can be sent wirelessly via the epoc Host and your LIS/ HIS to the patient EMR.
Hospital Quality Metrics
  • Improve patient experience and quality of care 
  • Reduce risk and increase compliance 
  • Standardize care 
Siemens Healthineers epoc Blood Analysis System
Quidel Triage MeterPro
Multi-Assay Instrument for BNP, Cardiac, D-Dimer, and Drugs of Abuse 
Perform a variety of cardiovascular, toxicology, and BNP tests at the point of care with the Quidel™ Triage™ MeterPro. It delivers results in 20 minutes and allows you to customize test panels via the Test Select™ feature. Use whole blood or EDTA plasma and urine specimens for drug screens. The BNP assay is CLIA-waived when used with whole blood.
Hospital Quality Metrics
  • Accelerate decision making for costly medical conditions
  • Support increased patient volumes in the ED
  • Shorten turnaround times from specimen collection to test result
Quidel Triage MeterPro
Roche Diagnostics CoaguChek XS System
Point-of-Care PT/INR Meter 
Get accurate fingerstick test results and the flexibility and control you need to confidently manage all of your anticoagulated patients with the CLIA-waived Roche Diagnostics CoaguChek™ XS System. It’s the only system that performs onboard quality control in the reagent chamber, and it delivers accurate and precise results in one minute. The meter neutralizes therapeutic levels of heparin and LMWH and is INR corrected for hematocrit within a specified range.
Hospital Quality Metrics
  • Confidently make clinical decisions with 97% correlation to lab results
  • Reduce heparin interference for more accurate results
Roche Diagnostics CoaguChek XS System