Amine and Photoreactive Crosslinkers
Thermo Scientific™ Pierce™ Sulfo-SANPAH Crosslinker
Crosslink amines to anything with this long crosslinker composed of Sulfo-NHS-ester and nitrophenyl azide reactive groups; photoactivates with UV light (Amax 320 to 350nm).
Thermo Scientific™ Pierce™ NHS-Diazirine Crosslinkers Available on GSA/VA Contract for Federal Government customers only.
Very short to mid-length (3.9 to 13.5A), amine-to-anything crosslinkers; NHS-ester and diazirine reactive groups; photoactivate with UV light (Amax 330 to 370nm).
  spinner