Vials
A variety of vials, closures, and vial accessories in a range of sizes, shapes, and material compositions designed for various scientific applications; includes autosampler vials and accessories, dilution vials, scintillation vials, and serum vials. Autosampler vials come in convenient pre-packaged sets.
Other options include general-purpose clear or amber glass and plastic sample vials and cryogenic storage vials for cell culture and biological samples.
Filtered Search Results
Fisherbrand™ Class B Clear Glass Threaded Vials with Closures Attached
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
General-purpose vial
| Disposable | Single-use |
|---|---|
| Type | Vial |
| Material (Liner) | Polyvinyl |
| Color | Clear |
| Classification | B |
| Material | Borosilicate Glass |
| Closure Type | Screw Cap |
| For Use With (Application) | For general purpose |
| Closure Material | Phenolic |
| Sterility | Non-sterile |
DWK Life Sciences™ Kimble™ Sample Vials with Black Phenolic Caps and Polyvinyl-faced Pulp Liners
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Kimble™ Screw thread N-51A Borosilicate Glass Sample Vials made of low extractable Kimax N-51A glass.
| Color | Clear |
|---|---|
| Closure Color | Black |
| Closure Material | Phenolic |
Fisherbrand™ Class B Clear Glass Threaded Vials
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
General purpose vial avoids alteration in pH of sample
| Disposable | Single-use |
|---|---|
| Type | Vial |
| Material (Liner) | Rubber |
| Color | Clear |
| Classification | B |
| Material | Borosilicate Glass |
| For Use With (Application) | For general purpose |
| Closure Material | Phenolic |
| Sterility | Non-sterile |
Fisherbrand™ Class A Clear Glass Threaded Vials with Attached Caps, PE Poly-Seal™ Cone Liner
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
With low-density polyethylene Poly-Seal cone liners
| Disposable | Single-use |
|---|---|
| Type | Vial |
| Material (Liner) | LDPE |
| Color | Clear |
| Classification | A |
| Certifications/Compliance | ASTM, USP Type I |
| For Use With (Application) | For general purpose |
| Closure Material | Phenolic |
| Sterility | Non-sterile |
| Product Line | Poly-Seal |
Fisherbrand™ Class A Clear Glass Threaded Vials
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
For general-purpose applications
| Thread Style | Screw |
|---|---|
| Disposable | Single-use |
| Material (Liner) | Rubber |
| Color | Clear |
| Classification | A |
| Material | Borosilicate Glass |
| Certifications/Compliance | ASTM E438, USP Type I, Type I |
| Autoclavable | Autoclavable |
| For Use With (Application) | For general purpose |
| Closure Material | Phenolic |
| Sterility | Non-sterile |
Qorpak™ Clear Borosilicate Sample Vials with Caps
Highly resistant to heat shock and chemical leaching
| Thread Style | Screw |
|---|---|
| Color | Clear |
| Material | Borosilicate Glass |
| Closure Type | Screw Cap |
| For Use With (Application) | Highly resistant to heat and chemicals |
Fisherbrand™ Class B Clear Glass Threaded Vials With Closures Packaged Separately
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
General-purpose vial
| Disposable | Single-use |
|---|---|
| Type | Vial |
| Material (Liner) | Polyvinyl |
| Color | Clear |
| Classification | B |
| Material | Borosilicate Glass |
| Certifications/Compliance | ASTM E438, USP Type I |
| Closure Type | Screw Cap (Closed-top) |
| For Use With (Application) | For general purpose |
| Closure Material | Phenolic |
| Sterility | Non-sterile |
DWK Life Sciences Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Manufactured from Wheaton 33 low extractable borosilicate glass. Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps are economically priced and conveniently packed in two shrink-wrapped packs which reduces contamination from corrugated material found in traditional packaging.
| Closure Type | Screw Cap |
|---|---|
| Autoclavable | Autoclavable |
Thermo Scientific™ Storage Vials
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
DWK Life Sciences Wheaton™ Clear Glass Sample Vials in Lab File with Caps Attached
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.
| Thread Style | Screw |
|---|---|
| For Use With (Equipment) | M-T File™ and Vial Rack |
| Color | Clear |
| Material | Borosilicate Glass (Low Extractable) |
| Certifications/Compliance | ASTM Type I, Class A, USP Type I |
| Closure Type | Screw Cap |
| Autoclavable | Autoclavable |
| For Use With (Application) | Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage |
| Closure Material | Phenolic |
Fisherbrand™ 9 mm Glass Screw Thread Vials
Fisherbrand™ 9 mm screw thread vials feature a wide neck opening which allows for easy filling. Available with marking spot and graduation marks for convenient sample identification.
| Height (Metric) | 32 mm |
|---|---|
| Volume (Metric) Total | 2 mL |
| Volume (Metric) Usable | 1.5 mL |
| Closure Size | 9 mm |
| Volume (Metric) | 1.5 mL |
| Material | Glass |
| Type | Screw Thread Autosampler Vial |
| Diameter (Metric) Outer | 12 mm |
| No. per Pack | 100 |
| Closure Type | Screw Cap |
| Volume (Metric) Residual | < 170 μL |
| Product Line | Fisherbrand |
| Base Style | Flat Bottom |
| Size (Metric) Opening | 9 mm |
Fisherbrand™ Borosilicate Glass Scintillation Vials, with White Polypropylene Caps
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Use these general purpose vials with all common counters. Fisherbrand™ 20mL Borosilicate Glass Scintillation Vials with white polypropylene caps feature potassium-free glass composition which provides consistently low activity counts and excellent light transmission.
| Height (Metric) | 61 mm |
|---|---|
| Volume (Metric) Total | 20 mL |
| Volume (English) | 0.67 oz. |
| Material (Liner) | Pulp-backed Foil |
| Closure Size | 22-400 |
| Packaging Type | Shrink-wrapped Tray |
| Color | White |
| Volume (Metric) | 20 mL |
| Material | Borosilicate Glass |
| Closure Material | Polypropylene |
| Type | Scintillation Vial |
| Marking Spot | Yes |
| Diameter (Metric) Outer | 28 mm |
| Diameter (English) Outer | 1.12 in. |
| Height (English) | 2.37 in. |
Fisherbrand™ Class B Amber Glass Threaded Vials with Attached Caps
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Type I, Class B amber borosilicate glass
| Type | Vial |
|---|---|
| Material (Liner) | Teflon-faced Rubber |
| Color | Amber |
| Classification | B |
| Material | Amber Glass (Class B) |
| Certifications/Compliance | ASTM, USP Type I |
| Closure Type | Fitted Cap |
| Autoclavable | Autoclavable |
| For Use With (Application) | For general purpose vials for light sensitive materials |
| Closure Material | Phenolic |
Fisherbrand™ Class B Amber Glass Threaded Vials
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
General-purpose vials for light-sensitive materials
| Disposable | Single-use |
|---|---|
| Type | Vial |
| Material (Liner) | Rubber |
| Color | Amber |
| Classification | B |
| Material | Borosilicate Glass |
| Closure Type | Screw Cap |
| Autoclavable | Autoclavable |
| For Use With (Application) | For general purpose vials for light sensitive materials |
| Closure Material | Phenolic |
| Sterility | Non-sterile |
| Includes | Hinged cap |
|---|---|
| Type | Vial |
| Material | LDPE,Polyethylene |
| For Use With (Application) | General purpose use |