SmartPack Chemical Packaging Solutions
- Overview
- Going Green
- EcoSafPak Packaging
- Pressure-Sensitive Safety Labels
- QR Codes
Our packaging enhancements provide a smarter, greener, and safer way to conduct research and transport products while helping to prevent costly accidents. Compliant with all applicable government regulations, our packaging solutions are designed to keep you safe, preserve the integrity of your chemicals, protect the environment, and provide convenient handling and storage.
Designed for Your Safety
Our Product Offerings Include:
Square Poly Bottles
Our award-winning recyclable polyethylene bottles are designed for safety and convenience, with a square shape that minimizes the storage space required for bottles.
Specialized Acid Containers
Fisher Chemical™ Optima™ Acids remain ultrapure in Teflon™ bottles that have been pretreated to minimize leaching and preserve the purity of contents.
Amber Glass Bottles
Designed to promote quality and reliability, amber glass bottles are used to package photosensitive chemicals to protect them from light.
Borosilicate Glass Bottles
Our borosilicate bottles significantly reduce contamination from metal cations (Na+ and K+) compared to soda-lime glass.
Plastic Bottles
Available when chemical properties are compatible, plastic bottles minimize the risk of breakage while providing lighter-weight packaging and easier, more economical shipping.
Aluminum Cans
With a lightweight, seamless construction that is designed to contain ethers, this option features a tamper-evident cap with pouring handle. They are pure aluminum, which minimizes Na+ and K+ adduct formation.
High-Volume Solvent Delivery Systems
We offer environmentally friendly solvent-handling solutions for virtually unlimited applications to enhance safety and improve productivity within your lab.
Going Green
Steps you can take to make your lab greener:
- Order your favorite Fisher Chemical products in EcoSafPak packaging manufactured by an SFI-certified manufacturer
- Consolidate orders or combine purchases with neighbor labs
- Order in cases instead of bottles, eliminating three times the packaging materials and lowering cost by one-third
- Utilize your local stockroom for small and urgent purchases to reduce packaging materials and CO2 emissions while avoiding expensive freight charges
EcoSafPak Packaging
Pressure-Sensitive Safety Labels
Fisher Chemical Labels Deliver CofA + SDS Via QR Codes
| Grade | Application |
|---|---|
| Optima UHPLC/MS | Ideal mobile phase for UHPLC-MS applications in pharmaceutical, biotechnology, clinical, environmental, and food safety industries |
| Optima LC/MS | Ideal mobile phase for LC/MS and UHPLC-UV applications in pharmaceutical, biotechnology, clinical, environmental, and food safety industries |
| Optima | HPLC, GC, plasma/ICP, spectrophotometry, and pesticide residue analysis |
| HPLC | HPLC and spectrophotometry procedures |
| Certified ACS | Analytical applications requiring tight specifications |
| Certified ACS Plus | Analytical applications with tighter metal specifications |
| Certified | General analytical procedures |
| Environmental Grade | HPLC, trace-organic analysis, environmental testing |
| GC Headspace | Performance tested to ensure absence of trace level ICH residual solvents |
| Pesticide | GC with electron capture detector (ECD), pesticide residue analysis |
| USP/NF/FCC/EP/BP/JP | Food and drug laboratories, biological testing |
| TraceMetal™ | Primarily used in digestion of samples prior to instrument (ICP) analysis |
| Laboratory, Technical, and Reagent | Manufacturing and general laboratory use |