Filtered Search Results
DuPont™ Tyvek™ 400 Extra Long Frocks, Front Snap Closure
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Tyvek frock extends to knee for applications that do not require the full body coverage of a coverall.
Disposable | Limited-use |
---|---|
Seam Type | Serged |
Cuff Style | Elastic |
Product Type | Extra Long Frock |
Color | White |
Style | Knee-length |
Material | Tyvek |
For Use With (Application) | Lead and asbestos abatement and remediation, general maintenance and operations, spray painting, general clean-up |
Closure Type | Snaps |
Collar Type | Laydown |
DuPont™ Tyvek™ 400 Coveralls with Elastic Wrists and Ankles, Vending Packaged
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Individually packaged coveralls for PPE vending feature elastic wrists and ankles to provide an ideal balance of protection, durability, and comfort.
Disposable | Limited-use |
---|---|
Neck Style | Collared |
Description | Flashspun HDPE |
Color | White |
Quantity | 25/Cs. |
Waist Style | Elastic |
Material | Tyvek |
Gender | Unisex |
For Use With (Application) | Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning |
Sterility | Non-sterile |
Resistance | Particle |
Seam Type | Serged |
Cuff Style | Elastic |
Product Type | General Purpose Coveralls |
Packaging | Individually Packaged for PPE Vending |
Model | TY125SWH |
Latex Free | Yes |
Ankle Style | Elastic |
Hood Type | Unhooded |
Certifications/Compliance | ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127 |
Closure Type | Front Zipper |
Collar Type | Laydown |
DuPont™ Tyvek™ 121S Series Coveralls, Elastic Wrists and Attached Boots
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Comfortable and durable flash spun high density polyethylene coveralls provides an inherent barrier against particles.
Disposable | Limited-use |
---|---|
Neck Style | Collared |
Color | White |
Material | Tyvek |
Gender | Unisex |
For Use With (Application) | Lead and Asbestos Abatement, Remediation, General Maintenance, Spray Painting, Cleaning |
Sterility | Non-sterile |
Seam Type | Serged |
Cuff Style | Elastic |
Product Type | General Purpose Coveralls |
Model | TY121S |
Ankle Style | Elastic,Attached Boots |
Hood Type | Unhooded |
Certifications/Compliance | USDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 |
Closure Type | Front Zipper |
Collar Type | Laydown |
DuPont™ Tyvek™ IsoClean™ Nonsterile Sleeves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Comfortable, lightweight and durable.
Disposable | Single-use |
---|---|
Length (Metric) | 45.7 cm |
Cuff Style | Elastic |
Product Type | Controlled Environment Sleeve |
Size | Standard |
Length (English) | 18 in. |
Model | IC501B |
Material | Tyvek |
DuPont™ Tyvek™ IsoClean™ Nonsterile Bouffants
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Creates a barrier to dry particles, microorganisms, and nonhazardous liquids. Suitable for ISO 5 (class 100) or higher cleanroom environments.
Includes | Elastic Headband |
---|---|
Disposable | Single-use |
Diameter (English) | 21.5 in. |
Seam Type | Serged |
Product Type | Cleanroom Bouffant |
Size | 21 in. |
Cleanroom Class | Class 10 (ISO 4) |
Color | White |
Quantity | 250/Cs. |
Material | Tyvek |
Certifications/Compliance | Category III PPE |
DuPont™ Tyvek™ IsoClean™ Series 447 Boot Covers
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Gripper™ soles resists slipping on slick surfaces
Height (Metric) | 45.7 cm |
---|---|
Seam Type | Serged |
Color | White |
Latex Free | Yes |
Material | Tyvek Flashspun Polyethylene |
For Use With (Application) | Ideal to provide an excellent barrier to dry particles, nonhazardous liquids and microorganisms |
Closure Type | Elastic Top and Ankles |
Height (English) | 18 in. |
DuPont™ Tyvek™ IsoClean™ Series 264 Frocks, Clean-Processed and Sterile
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Sterile and clean-processed frocks offer lowest level of particle shedding within the DuPont portfolio. Tyvek™ provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Disposable | Single-use |
---|---|
Seam Type | Serged |
Cuff Style | Elastic |
Product Type | Controlled Environments Frock |
Packaging Type | Clean-processed, Individually Packaged |
Color | White |
Sleeve Style | Raglan Sleeve |
Style | IsoClean™ 264 Series |
Material | Tyvek |
Closure Type | Zipper |
Collar Type | Bound |
Sterility | Sterile |
DuPont™ Tyvek™ IsoClean™ Zipper Front Cleanroom Frocks, Raglan Sleeves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bulk packaged in double transparent poly liners.
Seam Type | Serged |
---|---|
Cuff Style | Elastic |
Product Type | Controlled Environments Frock |
Packaging Type | Bulk Packaged |
Cleanroom Class | Class 1000 (ISO 6) |
Color | White |
Sleeve Style | Raglan Sleeve |
Material | Tyvek |
Closure Type | Zipper |
Collar Type | Bound |
Sterility | Non-sterile |
DuPont™ Tyvek™ IsoClean™ Boot Covers
Provides inherent barrier to dry particles, microorganisms, and non-hazardous liquids. Available in bulk or clean-processed packaging.
Height (Metric) | 46.99 cm |
---|---|
Sole Type | Anti-skid |
Cleanroom Class | Class 1000 (ISO 6) |
Color | White |
Latex Free | Yes |
Material | Tyvek Flashspun Polyethylene |
Certifications/Compliance | 16 CFR 1610 Class 1 |
Closure Type | Elastic Top with Ankle Ties |
Sterility | Non-sterile |
Height (English) | 18.5 in. |
DuPont™ Tyvek™ IsoClean™ Front Snap Cleanroom Frocks, High Mandarin Collar
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Cleanroom frocks feature a high mandarin collar with snap for a secure fit and provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Disposable | Single-use |
---|---|
Seam Type | Serged |
Cuff Style | Elastic |
Product Type | Controlled Environments Frock |
Cleanroom Class | Class 1000 (ISO 6) |
Color | White |
Sleeve Style | Set-in Sleeve |
Material | Tyvek |
Closure Type | Snaps |
Collar Type | High Mandarin |
Sterility | Non-sterile |
Neck Style | Hooded |
---|---|
Color | Yellow |
Material | Tychem |
For Use With (Application) | Petroleum refining, pulp and Paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing |
Closure Type | Zipper |
DuPont™ Tyvek™ Isoclean™ Series 255 Coveralls, Elastic Thumb Loops
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Disposable | Single-use |
---|---|
Seam Type | Serged |
Product Type | Cleanroom Coveralls |
Cleanroom Class | Class 1000 (ISO 6) |
Packaging | Bulk |
Sleeve Style | Dolman |
Hood Type | Standard |
Material | Tyvek™ Flashspun Polyolefin |
Certifications/Compliance | CE |
Gender | Unisex |
For Use With (Application) | Protective barrier to dry particles and microorganisms |
Sterility | Non-sterile |
DuPont™ Tyvek™ 400 Frocks, Open Wrists
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Tyvek frock features a body-length jacket design with front snap closures for easy donning and doffing. Available in individually packaged styles for PPE vending machines.
Disposable | Limited-use |
---|---|
Seam Type | Serged |
Cuff Style | Open |
Product Type | General Purpose Frock |
Color | White |
Material | Tyvek |
Certifications/Compliance | ASTM D1777 |
For Use With (Application) | Lead and asbestos abatement and remediation, general maintenance and operations, spray painting, general clean-up |
Closure Type | Snaps |
Collar Type | Laydown |
DuPont™ Tyvek™ 400 Coveralls, 125S Series, Elastic Wrists and Ankles
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Coveralls with elastic wrists and ankles provide an ideal balance of protection, durability, and comfort.
Disposable | Limited-use |
---|---|
Neck Style | Collared |
Description | Flashspun HDPE |
Color | White |
Quantity | 25/Cs. |
Waist Style | Elastic |
Material | Tyvek |
Gender | Unisex |
Sterility | Non-sterile |
Resistance | Particle |
Seam Type | Serged |
Cuff Style | Elastic |
Product Type | General Purpose Coveralls |
Latex Free | Yes |
Ankle Style | Elastic |
Hood Type | Unhooded |
Certifications/Compliance | ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127 |
Closure Type | Front Zipper |
Collar Type | Laydown |
DuPont™ Tyvek™ 400 Disposable Sleeve, NAFTA
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
DuPont™ Tyvek™ 400 disposable sleeves provide the protection and durability you need so you can focus on the task at hand.