Fisherbrand Entry Level Research Grade Inverted Microscope
Perfect for classroom observations of living cells and organisms under more natural conditions.
- Siedentopf head design for more flexible eyepiece adjustment
- Infinity-corrected optics for consistency between objectives
- Extra-long working distance (up to 195mm)
- Koehler LED illumination for uniform and glare-free viewing